Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATEH75DC01A8

AstaTech, Inc.

2-(2-CHLORO-4-NITROPHENYL)ACETONITRILE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
196.59
MDL number:

Iupac Name

(2-chloro-4-nitrophenyl)acetonitrile

Inchi Code

1S/C8H5ClN2O2/c9-8-5-7(11(12)13)2-1-6(8)3-4-10/h1-2,5H,3H2

InChI key

WLCJERYAJOGPSV-UHFFFAOYSA-N

Purity

80%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Sticky Oil to Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=172970

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=C90563


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service