Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z743691

ART® barrier tip reload inserts, ART essentials kit, 4 racks off ART 10 REACH, ART 200, ART 1000xl

Synonym(s):

aerosol barrier tips, filter tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
packaging:
pkg of 4X1152 ea
manufacturer/tradename:
Molecular BioProducts 2931-RI

packaging

pkg of 4X1152 ea

manufacturer/tradename

Molecular BioProducts 2931-RI

technique(s)

PCR: suitable

General description

Reload insert capable of holding 1152 tips, across 12 trays of 96 tips. Each insert contains 3 different tip types, there are 4 trays of each, ART 10 REACH, ART 200, ART 1000xl. Case of 4 reload inserts.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service