Documents
Z341878
IKA® universal attachments for KS 130/260 basic and control shakers
AS 130.1 universal attachment
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
packaging
pkg of 1 ea
manufacturer/tradename
IKA 8017300
General description
Note: This attachment can be adapted to all kinds of vessels by means of universal, infinitely variable clamping rolls.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG