Z262897
Wheaton narrow mouth bottles with caps
capacity 8 oz (250 mL), amber, white polypropylene cap, vinyl flat liner
Synonym(s):
Wheaton bottle, Wheaton bottles, narrow mouth
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
material
amber bottle
glass bottle
round bottle
vinyl flat liner
white polypropylene cap
joint
Joints threaded neck (24-400)
feature
with cap
packaging
pkg of 12 ea
manufacturer/tradename
WHEATON® W216839
capacity
8 oz (250 mL)
diam. × H
60 mm × 136 mm
color
amber
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Convenience packed with closures attached. Bottles made of Wheaton "900" amber glass or "800" clear soda lime flint glass.
Choice of cap styles:
Choice of cap styles:
- White PP cap with PTFE or polyvinyl liner
- Black plastic cap with Poly-Seal® PE cone liner
Features and Benefits
• Manufactured from amber, USP Type III soda-lime glass• The amber glass protects the contents from UV light• Polypropylene caps with choice of liners
Legal Information
Poly-Seal is a registered trademark of Wheaton Industries, Inc.
WHEATON is a registered trademark of Wheaton Industries, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service