Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W330205

Sigma-Aldrich

2-Methoxypyrazine

≥99%, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O
CAS Number:
Molecular Weight:
110.11
FEMA Number:
3302
EC Number:
Council of Europe no.:
11347
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.054
NACRES:
NA.21
organoleptic:
cocoa; nutty
grade:
FG
Halal
Kosher
biological source:
synthetic
agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 110

assay

≥99%

refractive index

n20/D 1.509 (lit.)

bp

60-61 °C/29 mmHg (lit.)

density

1.14 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

cocoa; nutty

SMILES string

COc1cnccn1

InChI

1S/C5H6N2O/c1-8-5-4-6-2-3-7-5/h2-4H,1H3

InChI key

WKSXRWSOSLGSTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

114.8 °F - closed cup

flash_point_c

46 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service