Documents
TEM-CFT300CU
Sigma-Aldrich
Carbon Film Supported Copper Thin Bar Square Mesh Grid
size 300 mesh, box of 25
Synonym(s):
CARBON FILM ONLY ON THIN BAR 300 MESH COPPER GRIDS 25/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 25
size
300 mesh
General description
A 5-6 nm thin film of pure carbon supported thin bar square mesh grid. Copper grid, 300 mesh. Carbon film show better electrical and heat conduction in the electron beam.