Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF47798139

Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti

foil, 150 x 150mm, 0.25mm thickness, annealed

Synonym(s):

FE210320

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

form

foil (annealed)

manufacturer/tradename

Goodfellow 477-981-39

size × thickness

150 mm × 150 mm × 0.25 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service