Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BMSE1101COMB1

Benchmark Extra Doublesided Combs for myGel ”InstaView” Casting Set

Synonym(s):

electrophoresis comb, gel comb

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.71

manufacturer/tradename

Benchmark Scientific E1101-COMB1

General description

Double sided comb, 22/12 teeth, for use with myGel Mini casting set

Application

myGel, casting, electrophoresis, agarose

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service