709514
1-Phenyl-4-penten-1-yne
97%
Synonym(s):
1-Phenylpent-4-en-1-yne, 5-Phenylpent-1-en-4-yne
About This Item
Recommended Products
assay
97%
form
liquid
density
0.934 g/mL at 25 °C
functional group
allyl
phenyl
SMILES string
C=CCC#Cc1ccccc1
InChI
1S/C11H10/c1-2-3-5-8-11-9-6-4-7-10-11/h2,4,6-7,9-10H,1,3H2
InChI key
XFUQCDNMEJGDIV-UHFFFAOYSA-N
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service