Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

689793

Sigma-Aldrich

(1S,2R)-trans-2-(Fmoc-aminomethyl)cyclohexanol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
CA$60.00
100 G
CA$125.00
500 G
CA$421.00

CA$60.00


Availability
Please contact Customer Service for Availability


Select a Size

Change View
25 G
CA$60.00
100 G
CA$125.00
500 G
CA$421.00

About This Item

Empirical Formula (Hill Notation):
C22H25NO3
CAS Number:
Molecular Weight:
351.44
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

CA$60.00


Availability
Please contact Customer Service for Availability

assay

≥96.5% (HPLC)
97%

form

powder

optical purity

enantiomeric excess: ≥97.5%

functional group

Fmoc

storage temp.

2-8°C

SMILES string

O[C@H]1CCCC[C@@H]1CNC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C22H25NO3/c24-21-12-6-1-7-15(21)13-23-22(25)26-14-20-18-10-4-2-8-16(18)17-9-3-5-11-19(17)20/h2-5,8-11,15,20-21,24H,1,6-7,12-14H2,(H,23,25)/t15-,21+/m1/s1

InChI key

DLDUSAROVXDNNQ-VFNWGFHPSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service