Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

653942

Sigma-Aldrich

(trans)-2-Cyclopropylvinylboronic acid pinacol ester

96%

Synonym(s):

(E)-2-Cyclopropylvinylboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19BO2
CAS Number:
Molecular Weight:
194.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

96%

refractive index

n20/D 1.466 (lit.)

bp

90-95 °C/3-4 mmHg (lit.)

density

0.937 g/mL at 25 °C (lit.)

SMILES string

CC1(C)OB(OC1(C)C)\C=C\C2CC2

InChI

1S/C11H19BO2/c1-10(2)11(3,4)14-12(13-10)8-7-9-5-6-9/h7-9H,5-6H2,1-4H3/b8-7+

InChI key

FCWYPDPDAZGFRJ-BQYQJAHWSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

199.9 °F

flash_point_c

93.3 °C

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service