Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

649945

Sigma-Aldrich

2-Bromo-3-pyridyl trifluoromethanesulfonate

96%

Synonym(s):

2-Bromo-3-pyridyl triflate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3BrF3NO3S
CAS Number:
Molecular Weight:
306.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

refractive index

n20/D 1.4806

density

1.7869 g/mL at 25 °C

SMILES string

BrC1=NC=CC=C1OS(=O)(C(F)(F)F)=O

InChI

1S/C6H3BrF3NO3S/c7-5-4(2-1-3-11-5)14-15(12,13)6(8,9)10/h1-3H

InChI key

DWJIUNKBHIJMPC-UHFFFAOYSA-N

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service