588067
Chloroethane-13C2
99 atom % 13C
Synonym(s):
Ethyl chloride-13C2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
isotopic purity
99 atom % 13C
bp
12.3 °C (lit.)
mass shift
M+2
SMILES string
[13CH3][13CH2]Cl
InChI
1S/C2H5Cl/c1-2-3/h2H2,1H3/i1+1,2+1
InChI key
HRYZWHHZPQKTII-ZDOIIHCHSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
signalword
Danger
Hazard Classifications
Aquatic Chronic 3 - Carc. 2 - Flam. Gas 1A - Press. Gas Liquefied gas - Repr. 1B
Storage Class
2A - Gases
wgk_germany
WGK 2
flash_point_f
-58.0 °F - closed cup
flash_point_c
-50.0 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service