Skip to Content
MilliporeSigma
All Photos(1)

Documents

497193

Sigma-Aldrich

1,4-Bis(hydroxydimethylsilyl)benzene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4[Si(CH3)2OH]2
CAS Number:
Molecular Weight:
226.42
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

95%

mp

133-136 °C (lit.)

SMILES string

C[Si](C)(O)c1ccc(cc1)[Si](C)(C)O

InChI

1S/C10H18O2Si2/c1-13(2,11)9-5-7-10(8-6-9)14(3,4)12/h5-8,11-12H,1-4H3

InChI key

YBNBOGKRCOCJHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A useful silicon nucleophile for Pd-catalyzed cross-coupling.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott E Denmark et al.
Journal of the American Chemical Society, 130(48), 16382-16393 (2008-11-13)
The sodium salts of allyldimethylsilanol and 2-butenyldimethylsilanol undergo palladium-catalyzed cross-coupling with a wide variety of aryl bromides to afford allylated and crotylated arenes. The coupling of both silanolates required extensive optimization to deliver the expected products in high yields. The
Denmark, S. E.; Ober, M. H.
Aldrichimica Acta, 36, 75-75 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service