463302
Glycolic acid ethoxylate 4-nonylphenyl ether
average Mn ~600
Synonym(s):
Nonoxynol-8 carboxylic acid
About This Item
Recommended Products
mol wt
average Mn ~600
impurities
≤1 wt. % NaCl
refractive index
n20/D 1.486 (lit.)
density
1.07 g/mL at 25 °C (lit.)
SMILES string
OCCO.OCC(O)=O.CCCCCCCCCc1ccc(O)cc1
InChI
1S/C15H24O.C2H4O3.C2H6O2/c1-2-3-4-5-6-7-8-9-14-10-12-15(16)13-11-14;3-1-2(4)5;3-1-2-4/h10-13,16H,2-9H2,1H3;3H,1H2,(H,4,5);3-4H,1-2H2
InChI key
HZXFKGPMHPPPMZ-UHFFFAOYSA-N
Application
All purpose cleaner.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service