Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

456934

Sigma-Aldrich

Dirhodium tetracaprolactamate

Synonym(s):

Tetrakis[μ-(hexahydro-2H-azepin-2-onato-κN1,κO2)]dirhodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H40N4O4Rh2
CAS Number:
Molecular Weight:
654.41
MDL number:
UNSPSC Code:
12352103

impurities

<2 mol/mol caprolactam

mp

269-274 °C (lit.)

SMILES string

[Rh][Rh].O=C1CCCCCN1.O=C2CCCCCN2.O=C3CCCCCN3.O=C4CCCCCN4

InChI

1S/4C6H11NO.2Rh/c4*8-6-4-2-1-3-5-7-6;;/h4*1-5H2,(H,7,8);;

Inchi Key

SSCZKSBMOZFVCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

For research purposes only-Not for Use in Humans or in Human Clinical Trials.
Under Special Agreement with Regis Technologies, Inc.

Legal Information

Sold under license from Regis Technologies, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service