Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

447730

Sigma-Aldrich

1,1,1,3,3,3-Hexakis(dimethylamino)diphosphazenium tetrafluoroborate

98%

Synonym(s):

Bis[tris(dimethylamino)phosphoranylidene]ammonium tetrafluoroborate, Phosphazenium tetrafluoroborate P2-BF4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[[(CH3)2N]3P=]2N(BF4)
CAS Number:
Molecular Weight:
427.21
Beilstein/REAXYS Number:
5470923
MDL number:
UNSPSC Code:
12352000

assay

98%

mp

≥260 °C (lit.)

SMILES string

F[B-](F)(F)F.CN(C)P(=[N+]=P(N(C)C)(N(C)C)N(C)C)(N(C)C)N(C)C

InChI

1S/C12H36N7P2.BF4/c1-14(2)20(15(3)4,16(5)6)13-21(17(7)8,18(9)10)19(11)12;2-1(3,4)5/h1-12H3;/q+1;-1

InChI key

DPIDWCQJGWFENO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Zer et al.
European journal of biochemistry, 231(2), 448-453 (1995-07-15)
The effect of 3-(3,4-dichlorophenyl)-1,1-dimethyl urea (diuron) binding at the secondary quinone (QB) binding site of reaction center II (RCII), on the high-light-induced degradation of the RCII proteins D1 and D2, and the core proteins CP43 and CP47 was investigated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service