Skip to Content
MilliporeSigma
All Photos(1)

Documents

443689

Sigma-Aldrich

3-Bromo-1,1,1-trifluoropropane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2CF3
CAS Number:
Molecular Weight:
176.96
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

refractive index

n20/D 1.363 (lit.)

bp

63.5 °C (lit.)

density

1.662 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)CCBr

General description

3-Bromo-1,1,1-trifluoropropane is a halogenated hydrocarbon.

Application

3-Bromo-1,1,1-trifluoropropane may be used for the synthesis of 3-(benzyloxy)-N-3,3,3-trifluoropropyl-16,17-seco-estra-1,3,5(10)-triene-16,17-imide and [N-hydroxy-N-(1-trifluoromethylethenyl)]amido
diethylphosphate.
Please view www.aldrich.com/epaods regarding the EPA′s request for application information of Ozone Depleting Substances

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Ozone 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphorus substituted hydroxylamine and hydroxamic acid derivatives: synthesis and reactivity.
Alonso C, et al.
ARKIVOC (Gainesville, FL, United States), 221-253 (2011)
Y Tanaka et al.
Archives of biochemistry and biophysics, 263(1), 178-190 (1988-05-15)
Leukotriene B4 is rapidly metabolized through omega-oxidation, preventing its detection when it is produced under certain biological conditions. To investigate leukotriene B4 production in various physiological conditions, analogs of arachidonic acid which are converted to metabolically stable analogs of leukotriene
L W Lawrence Woo et al.
Molecular cancer therapeutics, 7(8), 2435-2444 (2008-08-30)
An improved steroid sulfatase inhibitor was prepared by replacing the N-propyl group of the second-generation steroid-like inhibitor (2) with a N-3,3,3-trifluoropropyl group to give (10). This compound is 5-fold more potent in vitro, completely inhibits rat liver steroid sulfatase activity
Tetrahedron Letters, 37, 5557-5557 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service