Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

422576

Sigma-Aldrich

3-Benzoylpyridine-2-carboxylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9NO3
CAS Number:
Molecular Weight:
227.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

powder

mp

151-155 °C (lit.)

functional group

carboxylic acid
ketone
phenyl

SMILES string

OC(=O)c1ncccc1C(=O)c2ccccc2

InChI

1S/C13H9NO3/c15-12(9-5-2-1-3-6-9)10-7-4-8-14-11(10)13(16)17/h1-8H,(H,16,17)

InChI key

YERHKEWRHQIXFY-UHFFFAOYSA-N

General description

3-Benzoylpyridine-2-carboxylic acid is a pyridine derivative. It is a heterocyclic building block used in chemical synthesis.

Application

3-Benzoylpyridine-2-carboxylic acid may be used in the synthesis of 5-methyl-N-aryl substituted-1,3-oxazolidine-2,4-dione derivatives.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and antitubercular activity of 5-methyl-N-aryl substituted oxazolidine dione derivatives.
International Journal of Pharmaceutical Archive, 2(5), 95-102 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service