Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

408409

Sigma-Aldrich

trans-N-(2-Furfurylideneacetyl)glycine

98%

Synonym(s):

N-[(2E)-3-(2-Furanyl)-1-oxo-2-propen-1-yl]-glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO4
CAS Number:
Molecular Weight:
195.17
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

98%

mp

218-221 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)CNC(=O)\C=C\c1ccco1

InChI

1S/C9H9NO4/c11-8(10-6-9(12)13)4-3-7-2-1-5-14-7/h1-5H,6H2,(H,10,11)(H,12,13)/b4-3+

Inchi Key

XJVSWKBQMAOKAX-ONEGZZNKSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Kanstrup et al.
Analytical biochemistry, 194(1), 41-44 (1991-04-01)
Furylacryloyl substrates used in kinetic measurements of proteolytic enzymes are shown to cis-trans photoisomerize quickly in plain daylight. The enzymatic transformation of the two forms is sufficiently different to cause problems for such measurements without careful protection against light.
M K Parkash et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 32(10), 887-895 (1994-10-01)
The fate of furfural (2-furancarboxaldehyde) was investigated in male and female Fischer 344 (F344) rats given single oral doses of 1, 10 and 60 mg/kg and male and female CD1 mice given 1, 20 and 200 mg/kg [carbonyl-14C]furfural. There was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service