Skip to Content
MilliporeSigma
All Photos(1)

Documents

181862

Sigma-Aldrich

Polyepichlorohydrin

average Mw ~700,000 by GPC

Synonym(s):

Epichlorohydrin homopolymer, Epoxychloropropane homopolymer, Poly[(chloromethyl)ethylene oxide]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH(CH2Cl)CH2O]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

slab/chunk

mol wt

average Mw ~700,000 by GPC

density

1.36 g/mL at 25 °C (lit.)

SMILES string

O\C=C/CCl

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service