Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

161608

Sigma-Aldrich

2-Ethylcyclohexanol, mixture of cis and trans

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H10OH
CAS Number:
Molecular Weight:
128.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

form

solid

refractive index

n20/D 1.464 (lit.)

bp

74-79 °C/12 mmHg (lit.)

density

0.906 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCC1CCCCC1O

InChI

1S/C8H16O/c1-2-7-5-3-4-6-8(7)9/h7-9H,2-6H2,1H3

InChI key

CFYUBZHJDXXXQE-UHFFFAOYSA-N

General description

2-Ethylcyclohexanol exhibits self-association due to larger flexibility of the cyclohexanol ring in inert solvents[1].

Application

2-Ethylcyclohexanol was used in the preparation of cyclohexanone[2].

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

154.4 °F - closed cup

flash_point_c

68 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of microbiologically and enzymatically mediated Baeyer-Villiger oxidations: synthesis of optically active caprolactones.
Alphand V, et al.
Journal of the Chemical Society. Perkin Transactions 1, 15, 1867-1872 (1996)
Self-association of cyclohexanols in inert solvents. Apparent heat capacities of cyclohexanol and substituted cyclohexanols in n-heptane and n-decane.
Trejo LM, et al.
J. Chem. Soc., Faraday, 87(11), 17393-17343 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service