AATB-23010
AAT Bioquest
AAT Bioquest Cell Meter™ Cell Adhesion Assay Kit
100 Tests
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
fluorescence
λex 494 nm;λem 514 nm
manufacturer/tradename
AAT Bioquest
General description
The Cell Meter™ Cell Adhesion Assay Kit is a fast and sensitive assay for measuring cell-cell or cell-surface adhesion for a variety of cell types. In this assay, cells are labeled with Calcein UltraGreen AM and allowed to adhere. After removal of nonadherent cells, The fluorescence of Calcein UltraGreen is used to calculate the number of adherent cells.
Components:
- Adhesion Assay Buffer-1 bottle (25 mL)
- Calcein UltraGreen™ AM-1 vial
- DMSO-1 vial (100 µL)
Legal Information
Cell Meter™ is a trademark of AAT Bioquest
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.