MilliporeSigma
mAbs Formulation
and Fill Finish
Products for Development and Manufacturing
Formulating, sterile filtering, and filling a drug product is complex but crucial. There are many excipient choices, filter options, and fill-finish technologies, each of which can have an impact on key attributes of the final dosage form including stability.
Final Sterile Filtration & Filling
Final sterilizing filtration or bulk filtration is a critical step in the monoclonal antibody (mAb) manufacturing processes as it ensures bacterial contaminants are removed from the drug product. Multiple regulatory guidance documents describe specific considerations for filtration operations, including requirements for filter integrity testing and expectations for establishing compatibility of the filtration system with the process stream.
Final filling involves transfer of sterile filtered drug product to vials, syringes, or other containers in a highly controlled filling environment, designed to maintain sterility and mitigate any risk of product contamination.
We can support you in making informed choices for your filtration and filling operation with products and services tailored to this critical process step.
  • Millipore® Filters
  • Millipore® Single-Use Solutions (2D/3D single-use assemblies and storage systems, final fill assemblies, connectors)
  • Emprove® Dossiers to Support Process Optimization and Safety Risk Assessment with Detailed Extractables Profile
  • Millipore® Equipment & Systems (integrity tester, housings)
  • Millipore® Sampling Solution (offline system)
  • Millipore® Services (single-use and filter validation, system services, equipment installation, system qualification, system training)
  • Millipore® Automation and Analytics Software
  • BioReliance® Final Product Release Testing
  • Millipore® CTDMO Services
Related Materials
Loading
Related Categories
Parenteral formulations using protein stabilizers
Protein Stabilizers
Our portfolio of pharmaceutical excipients offers everything you need for medicinal drug product.
Learn More
Reducing viscosity of highly concentrated monoclonal antibody (mAb) formulation to enable subcutaneous delivery
Viscosity Reduction Platform
Our Viscosity Reduction Platform enables subcutaneous formulation of highly concentrated proteins.
Learn More
Biopharma Formulation
Biopharmaceutical Formulation
We offer low bioburden, endotoxin materials for biopharma formulation.
Learn More
Integritest® 5 automated test instrument used to test the integrity of filters or processing equipment
Integrity Testers
The Integritest® 5 automated test instrument enables non-destructive integrity testing to be performed inline or offline on filtration devices.
Learn More
Mobius formulation fill
Final Sterile Filtration & Filling
Mobius® single-use systems can be designed to include either single or redundant sterilizing filters.
Learn More
Mobius® 3D Bag Assembly in 3D Large Liquid Transport Bin
Fluid Management
Robust fluid management systems mitigate the risk of process contamination. By combining our portfolio of single-use and multi-use systems, we can support your process needs.
Learn More
Related Resources
Page 1 of 2
Explore Our Solutions