CLS8513
Corning® storage tube racks
Synonym(s):
384 tube storage racks, empty storage racks, storage racks
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122804
NACRES:
NB.13
Recommended Products
material
polypropylene
sterility
non-sterile
feature
Holds 384 tubes
packaging
pack of 1
case of 20
manufacturer/tradename
Corning 8513
size
384 wells
color
blue
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service