Skip to Content
Merck
All Photos(1)

Documents

NI 4 426/7A3-3

from Mouse x Mouse Hybridoma, 92030618, Reactivity Anti-interleukin-4, human )

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

NS0

Reactivity

Anti-interleukin-4, human

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

Cell Line Origin

Mouse NS0 x Mouse BALB/c spleen

Cell Line Description

This neutralising antibody binds to all forms of human IL-4 tested, natural and recombinant.

Immunogen

Recombinant human interleukin-4

Application

Immunoassays

Culture Medium

RPMI 1640 + 2 mM glutamine + 10% Fetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: All customers are required to complete a Cell line Release Authorisation form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service