Skip to Content
Merck
All Photos(3)

Key Documents

MABT100

Sigma-Aldrich

Anti-Girdin Antibody, CT, clone 10E6.1

clone 10E6.1, from mouse

Synonym(s):

Girdin, Akt phosphorylation enhancer, Coiled-coil domain-containing protein 88A, G alpha-interacting vesicle-associated protein, GIV, Girders of actin filament, Hook-related protein 1, HkRP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
10E6.1, monoclonal
application:
ICC
IHC
IP
WB
species reactivity:
human
technique(s):
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable
citations:
3

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10E6.1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CCDC88A(55704)

General description

Girdin (Ccdc88a; APE, GIV, or HkRP1) is a ubiquitous protein belonging to the CCDC88 family. Girdin is an actin-binding protein which interacts with actin via its C-terminal domain, and may play a crucial role in cell motility and cytoskeleton remodeling. These effects may be regulated by a reciprocal relationship between Girdin and Akt. Akt phosphorylates Girdin, and Girdin binds to Akt and enhances phosphorylation of Akt, thereby promoting phosphorylation of Akt’s downstream targets such as GSK-3β and FOXO1A. Girdin/Akt signaling may play an important role in cancer by facilitating the proliferation of cancer cells.

Immunogen

Recombinant protein corresponding to human Girdin.

Application

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Girdin in epithelial cells of human prostate tissue.

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected Girdin in HeLa cells.

Immunoprecipitation Analysis: A representative lot immunoprecipitated Girdin in COS-7 cell lysate.
This Anti-Girdin, C-terminus Antibody, clone 10E6.1 is validated for use in Western Blotting, IHC(P), ICC, Immunoprecipitation for the detection of Girdin, C-terminus, clone 10E6.1.

Quality

Evaluated by Western Blot in human brain tissue lysate.

Wesetern Blot Analysis: A 1:5000 dilution of this antibody detected Girdin in 10 µg of human brain tissue lysate.

Target description

~216 kDa observed

Physical form

Format: Purified

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keisuke Taniuchi et al.
Oncotarget, 10(30), 2869-2886 (2019-05-14)
We report the use of small interfering RNAs (siRNAs) against ARHGEF4, CCDC88A, LAMTOR2, mTOR, NUP85, and WASF2 and folic acid (FA)-modified polyethylene glycol (PEG)-chitosan oligosaccharide lactate (COL) nanoparticles for targeting, imaging, delivery, gene silencing, and inhibition of invasiveness and metastasis
Aki Tanouchi et al.
Journal of experimental & clinical cancer research : CR, 35(1), 190-190 (2016-12-07)
Coiled-Coil Domain Containing 88A (CCDC88A) was identified as a substrate of the serine/threonine kinase Akt that is capable of binding to the actin cytoskeleton. The aim of this study was to investigate the potential role of CCDC88A in the migration
Pradipta Ghosh et al.
Current protocols in chemical biology, 8(4), 265-298 (2016-12-08)
Canonical signal transduction via heterotrimeric G proteins is spatiotemporally restricted, i.e., triggered exclusively at the plasma membrane, only by agonist activation of G protein-coupled receptors via a finite process that is terminated within a few hundred milliseconds. Recently, a rapidly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service