475986
mTOR Kinase Inhibitor II, WYE-354
The mTOR Kinase Inhibitor II, WYE-354, also referenced under CAS 1062169-56-5, controls the biological activity of mTOR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
mTOR Kinase Inhibitor II, WYE-354, Methyl-4-(6-{4-[(methoxycarbonyl)amino]phenyl}-4-morpholin-4-yl-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidine-1-carboxylate, mTOR Inhibitor II
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
DMSO: 50 mg/mL
shipped in
ambient
storage temp.
−20°C
SMILES string
N5(CCOCC5)c1nc(nc3c1C=[N]#[N]3C4CCN(CC4)C(=O)OC)c2ccc(cc2)NC(=O)OC
InChI
1S/C24H29N7O5/c1-34-23(32)26-17-5-3-16(4-6-17)20-27-21(29-11-13-36-14-12-29)19-15-25-31(22(19)28-20)18-7-9-30(10-8-18)24(33)35-2/h3-6,15,18H,7-14H2,1-2H3,(H,26,32)
InChI key
IMXHGCRIEAKIBU-UHFFFAOYSA-N
General description
Packaging
Warning
Reconstitution
Other Notes
Zask, A., et al. 2009. J. Med. Chem.52, 5013.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service