Skip to Content
Merck
All Photos(1)

Key Documents

203694

Sigma-Aldrich

bpV(bipy)

A potent inhibitor of phosphotyrosine phosphatase (Ki = 100 nM for insulin receptor dephosphorylation).

Synonym(s):

bpV(bipy), Potassium Bisperoxo(bipyridine)oxovanadate (V), PTEN Inhibitor II, PTP Inhibitors XIV

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K[VO(O2)2C10H8N2] · 3H2O
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (51V-NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow to orange

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A potent inhibitor of phosphotyrosine phosphatase activity (Ki = 100 nM for insulin receptor dephosphorylation). An activator of insulin receptor kinase (IRK). Also reported to be a potent inhibitor of PTEN (IC50 = 18 nM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
phosphotyrosine phosphatase
Product does not compete with ATP.
Reversible: no
Target IC50: 18 nM against PTEN
Target Ki: 100 nM for phosphotyrosine phosphatase

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Other Notes

Schmid, A.C., et al. 2004. FEBS Lett.566, 35.
Posner, B.I., et al. 1994. J. Biol. Chem. 269, 4596.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service