Skip to Content
Merck
All Photos(1)

Key Documents

1.06505

Supelco

Sodium dithionite

EMPLURA®

Synonym(s):

Sodium dithionite, Sodium hydrosulfite, Sodium hypodisulfite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2O4S2
CAS Number:
Molecular Weight:
174.11
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-890-0
NACRES:
NA.21
form:
powder
solubility:
250 g/L (slow decomposition)

Quality Level

product line

EMPLURA®

Assay

≥85.0% (iodometric)

form

powder

autoignition temp.

>200 °C

potency

2500 mg/kg LD50, oral (Rat)

pH

5.5-8.5 (20 °C, 50 g/L in H2O)

mp

100 °C (decomposition)

transition temp

flash point >100 °C

solubility

250 g/L (slow decomposition)

density

2.5 g/cm3 at 20 °C

bulk density

1250 kg/m3

anion traces

chloride (Cl-): ≤0.05%

cation traces

Fe: ≤0.005%

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[S](=O)([S](=O)[O-])[O-]

InChI

1S/2Na.H2O4S2/c;;1-5(2)6(3)4/h;;(H,1,2)(H,3,4)/q2*+1;/p-2

InChI key

JVBXVOWTABLYPX-UHFFFAOYSA-L

Application


  • Fast detection of sodium dithionite in sugar using a xanthylium-based fluorescent probe.: This study introduces a rapid method for detecting sodium dithionite residues in sugar using a novel xanthylium-based fluorescent probe. The method is highly sensitive and efficient, making it a significant advancement in food safety and quality control (Wang et al., 2024).

  • Stabilising mannose using sodium dithionite at alkaline conditions.: Research in the kraft pulping process explores the use of sodium dithionite to stabilize mannose under alkaline conditions. The findings highlight its potential to reduce hemicellulose degradation, providing insights for enhancing the efficiency of chemical pulping processes (Lindén et al., 2020).

Analysis Note

Assay (iodometric): ≥ 85.0 %
Identity: passes test
Chloride (Cl): ≤ 0.05 %
Fe (Iron): ≤ 0.005 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Self-heat. 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

212.0 °F

Flash Point(C)

> 100 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service