Skip to Content
Merck
All Photos(1)

Key Documents

1.03067

Supelco

N,N-Dimethyl-1,4-phenylenediammonium dichloride

GR for analysis

Synonym(s):

N,N-Dimethyl-1,4-phenylenediammonium dichloride, 4-Amino-N,N-dimethylaniline dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2 · 2HCl
CAS Number:
Molecular Weight:
209.12
MDL number:
UNSPSC Code:
12352107
EC Index Number:
208-635-7
NACRES:
NA.05

Quality Level

Assay

≥99.5% (acidimetric)

form

solid

pH

1.6 (20 °C, 50 g/L in H2O)

mp

208-212 °C (decomposition)

solubility

2000 g/L

bulk density

500 kg/m3

storage temp.

2-30°C

InChI

1S/C8H12N2.2ClH/c1-9-7-3-5-8(10-2)6-4-7;;/h3-6,9-10H,1-2H3;2*1H

InChI key

PXJHVKRLFWZUNV-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 99.5 %
Identity (IR-spectrum): passes test
Sulphate (SO₄): ≤ 0.05 %
Sulfated ash: ≤ 0.05 %

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service