OTV000364
8-[(Diisobutylamino)methyl]-7-hydroxy-3-(4-propylphenoxy)-2-(trifluoromethyl)-4H-chromen-4-one
AldrichCPR
About This Item
form
solid
SMILES string
FC(F)(F)C1=C(C(=O)c3c(c(c(cc3)[O-])C[N+H](CC(C)C)CC(C)C)O1)Oc2ccc(cc2)CCC
InChI
1S/C28H34F3NO4/c1-6-7-19-8-10-20(11-9-19)35-26-24(34)21-12-13-23(33)22(25(21)36-27(26)28(29,30)31)16-32(14-17(2)3)15-18(4)5/h8-13,17-18,33H,6-7,14-16H2,1-5H3
InChI key
YKNMOOSOMQCHJZ-UHFFFAOYSA-N
Other Notes
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service