DWKCPB2000P
Azlon Hexagonal Base Polypropylene Cylinders with Printed Graduations
class B, capacity 2000 mL , with +/20mL tolerance
Synonym(s):
CPB2000P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
class B
graduated
packaging
pkg of 1 ea
manufacturer/tradename
Azlon CPB2000P
capacity
2000 mL , with +/20mL tolerance
Looking for similar products? Visit Product Comparison Guide
Application
Suitable for the measurement and dispensing of liquids in the laboratory
Features and Benefits
Conform to ISO 6706 with anti-suction designto prevent base sticking to wet surface
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service