452815
Trimethoxy(7-octen-1-yl)silane
technical grade, 80%
Synonym(s):
(7-Octen-1-yl)trimethoxysilane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
H2C=CH(CH2)6Si(OCH3)3
CAS Number:
Molecular Weight:
232.39
Beilstein:
2437131
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
grade
technical grade
Assay
80%
refractive index
n20/D 1.427 (lit.)
bp
48-49 °C/0.1 mmHg (lit.)
density
0.928 g/mL at 25 °C (lit.)
SMILES string
CO[Si](CCCCCCC=C)(OC)OC
InChI
1S/C11H24O3Si/c1-5-6-7-8-9-10-11-15(12-2,13-3)14-4/h5H,1,6-11H2,2-4H3
InChI key
RKLXSINPXIQKIB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Trimethoxy(7-octen-1-yl)silane was used to silanize the surface of coverslips which were then used for DNA combing. The silane can be used as a coupling agent to link methacrylate-based polymers to a siliceous matrix.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
201.2 °F - closed cup
Flash Point(C)
94 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Tom Morrison et al.
Nucleic acids research, 34(18), e123-e123 (2006-09-27)
Understanding biological complexity arising from patterns of gene expression requires accurate and precise measurement of RNA levels across large numbers of genes simultaneously. Real time PCR (RT-PCR) in a microtiter plate is the preferred method for quantitative transcriptional analysis but
Preparation and characterization of mesostructured polymer-functionalized sol?gel-derived thin films.
Progress in Organic Coatings, 47(2), 393-400 (2003)
Karin Koch et al.
Physical chemistry chemical physics : PCCP, 22(4), 2087-2097 (2020-01-07)
Switching of liquid crystal phases is of enormous technological importance and enables digital displays, thermometers and sensors. As an alternative to electric fields or temperature, magnetic fields are an interesting trigger, as they are on the one hand versatile to
Silanization of coverslips for DNA combing.
Biotechniques, 11, 50-50 (2009)
Hui-Jiuan Chen et al.
Scientific reports, 8(1), 12600-12600 (2018-08-24)
Many fields of applications require dispersion of hydrophobic particles in water, which is traditionally achieved by using surfactants or amphiphilic molecules to modify particle surfaces. However, surfactants or amphiphilic molecules may disturb the native solution or particles' surface hydrophobicity, limiting
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service