Skip to Content
Merck
All Photos(1)

Key Documents

246115

Sigma-Aldrich

4,5-Imidazoledicarboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N2O4
CAS Number:
Molecular Weight:
156.10
Beilstein:
147774
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

97%

form

powder

mp

280-285 °C (dec.) (lit.)

SMILES string

OC(=O)c1nc[nH]c1C(O)=O

InChI

1S/C5H4N2O4/c8-4(9)2-3(5(10)11)7-1-6-2/h1H,(H,6,7)(H,8,9)(H,10,11)

InChI key

ZEVWQFWTGHFIDH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xing Yang et al.
Contrast media & molecular imaging, 11(4), 304-312 (2016-04-14)
A series of intra-molecular hydrogen bonded imidazoles and related heterocyclic compounds were screened for their N-H chemical exchange saturation transfer (CEST) magnetic resonance imaging (MRI) contrast properties. Of the compounds, imidazole-4,5-dicarboxamides (I45DCs) were found to provide the strongest contrast, with
Ali Behbahani et al.
Journal of AOAC International, 98(5), 1417-1422 (2015-11-04)
A new method was applied to produce modified nanoporous silica as a novel sorbent for Pb(II) ion SPE from industrial wastewater samples. In this modified method, the produced nanoporous silica has a higher functional group loading, which leads to a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service