All Photos(1)
About This Item
Linear Formula:
Cl2C6H3NHNH2 · HCl
CAS Number:
Molecular Weight:
213.49
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
95%
form
powder
mp
208-210 °C (dec.) (lit.)
functional group
chloro
hydrazine
SMILES string
Cl.NNc1cc(Cl)cc(Cl)c1
InChI
1S/C6H6Cl2N2.ClH/c7-4-1-5(8)3-6(2-4)10-9;/h1-3,10H,9H2;1H
InChI key
BDXZOJVMTJOAPS-UHFFFAOYSA-N
Application
3,5-Dichlorophenylhydrazine hydrochloride was used to prepare [2-(3,5-dichlorophenyl)-1,1-dioxo-2,3-dihydro-1H-1λ6-[1,2,3,5]thiatriazol-4-yl]dimethylamine.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Peter J Duggan et al.
Organic & biomolecular chemistry, 5(3), 472-477 (2007-01-26)
An unusual ring-expansion reaction of 4-amino-1,1-dioxo-[1,2,3,5]-thiatriazoles has been identified that produces the relatively rare 5-amino-1,1-dioxo-[1,2,4,6]-thiatriazines and. Initial alkylation of the thiatriazole with alpha-halo-esters at N-3 produces alpha-substituted esters which, under basic reaction conditions, undergo opening of the thiatriazole ring and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service