Skip to Content
Merck
All Photos(1)

Key Documents

139750

Sigma-Aldrich

4-(Dodecyloxy)benzoic acid

98%

Synonym(s):

p -(Dodecyloxy)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OC6H4CO2H
CAS Number:
Molecular Weight:
306.44
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid crystal

SMILES string

CCCCCCCCCCCCOc1ccc(cc1)C(O)=O

InChI

1S/C19H30O3/c1-2-3-4-5-6-7-8-9-10-11-16-22-18-14-12-17(13-15-18)19(20)21/h12-15H,2-11,16H2,1H3,(H,20,21)

InChI key

ALQLYJHDBAKLBB-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Occurrence of Ambient Temperature and Re-Entrant Smectic Ordering in an Inter Molecular Hydrogen Bonding between Alkyl Aniline and Alkoxy Benzoic Acids
Chitravel, T., and MLN Madhu Mohan
Bulgarian Journal of Physics, 36(4), 274-285 (2009)
Determination of the self-association and inter-association equilibrium constants of a carboxylic acid and its mixtures with pyridine derivates
Gonzalez, A., et al.
Vibrational Spectroscopy, 41(1), 21-27 (2006)
Formation of columnar hexagonal mesophases near room temperature from functionalised [9] aneNS2 (1, 4-dithia-7-azacyclononane)
Blake, Alexander J., et al.
Journal of Materials Chemistry, 11(4), 1011-1018 (2001)
Induced crystal G Phase through intermolecular hydrogen bonding in nOBA: 9HB
S. Sriharisastry, C.Nageswara Rao, T. Vishwam, K. Mallika, B. Gowrishankar Rao, Ha Sie Tiong.
Research Journal of Physical Sciences, 1(6), 1-10 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service