Przejdź do
58721
Tubing Adapter
1/4-28 male UNF, female Luer
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
218,00 zł
Informacje o tej pozycji
UNSPSC Code:
41121815
NACRES:
NB.21
eCl@ss:
32060190
218,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Złóż zamówienie na produkt na zamówienie
Pomoc techniczna
Pozwól nam pomóc
material
Kel-F (CTFE)
description
for chromatography applications
packaging
pkg of 1 ea
manufacturer/tradename
VICI Valco® CFLAPFA
fitting
female Luer, 1/4-28 male UNF
General description
Female adapter (1/16" I.D.) connects syringe to ¼"-28 threaded tubing or adapter.
Tubing adaptor is a national pipe thread taper made up of stainless steel which is used to connect a female adaptor. It may be used to connect capillary tubes in HPLC, GC etc.
Application
Tubing adaptor was used in solid phase extraction.
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
587225874358742
Tubing Adapter 1/4-28 male UNF, female Luer
58721
Tubing Adapter
Tubing Adapter 1/4-28 male UNF, male Luer
58722
Tubing Adapter
Tubing Adapter 1/4-28 male UNF, for 1/8 in. tube
58743
Tubing Adapter
Tubing Adapter 1/4-28 male UNF, for 1/4 in. O.D. tube
58742
Tubing Adapter
description
1/4-28 male UNF, female Luer
description
1/4-28 male UNF, male Luer
description
1/4-28 male UNF, for 1/8 in. tube
description
1/4-28 male UNF, for 1/4 in. O.D. tube
fitting
1/4-28 male UNF, female Luer
fitting
1/4-28 male UNF, male Luer
fitting
for 1/8 in. tube, 1/4-28 male UNF
fitting
1/4-28 male UNF, for 1/4 in. O.D. tube
manufacturer/tradename
VICI Valco® CFLAPFA
manufacturer/tradename
VICI Valco® CMLAKF
manufacturer/tradename
-
manufacturer/tradename
-
material
Kel-F (CTFE)
material
CTFE
material
stainless steel unit
material
CTFE unit
description
for chromatography applications
description
for chromatography applications
description
for chromatography applications
description
for chromatography applications
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
Still not finding the right product?
Explore all of our products under Tubing Adapter
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)