Merck
58289
Waters® Union
size 1/16 in., stainless steel
Informacje o tej pozycji
UNSPSC Code:
41115700
NACRES:
NB.21
Pomoc techniczna
Pozwól nam pomóc
material
stainless steel
packaging
pkg of 1 ea
size
1/16 in.
General description
Waters® Union is used for connecting most HPLC columns to a 1/16 in. tubing system.
Other Notes
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Waters is a registered trademark of Waters
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)