Przejdź do zawartości
58283
Tee
zero dead volume, for connecting 1/16 in., bore size 0.75 mm
Synonim(y):
Valco® Unions
Informacje o tej pozycji
UNSPSC Code:
41115700
NACRES:
NB.21
Pomoc techniczna
Pozwól nam pomóc
packaging
pkg of 1 ea
manufacturer/tradename
VICI Valco® ZT1
bore size
0.75 mm
compatibility
for connecting 1/16 in.
General description
Stainless steel, unless noted otherwise. Nuts and ferrules included.
Other Notes
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Still not finding the right product?
Explore all of our products under Tee
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)