Przejdź do zawartości
57653
Upchurch PEEK LiteTouch® Fitting
double-winged natural PEEK nut
Informacje o tej pozycji
UNSPSC Code:
41115700
NACRES:
NB.21
Pomoc techniczna
Pozwól nam pomóc
material
double-winged natural PEEK nut
description
litetouch fitting
packaging
pkg of 10 ea
fitting
1/4-28 male unit for 1/8 in. O.D. tubing (requires LiteTouch ferrule)
General description
Unique design prevents twisting of PEEK tubing. The fitting consists of a special PEEK ferrule, a stainless steel ring, and a PEEK or stainless steel nut for 1/16 in. tubing. The fitting will not leak at pressures to 5000 psi/340 bar with the PEEK nut, 2500 psi/175 bar with the stainless steel nut finger-tightened or 7000 psi/480 bar with wrench tightening. After wrench-tightening the stainless steel nut, subsequent finger-tightening will hold to 5000 psi. Compatible with all manufacturers′ stainless steel nuts. Order nuts and ferrules separately.
Other Notes
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
LiteTouch is a registered trademark of IDEX Health & Science, LLC
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)