Merck
57632
Sample Loop for Model 7725 Injector
volume 5 mL
Informacje o tej pozycji
UNSPSC Code:
41115854
NACRES:
NB.21
Pomoc techniczna
Pozwól nam pomóc
volume
5 mL
General description
Sample loop for model 7725 injector (volume 5 μL) is fabricated for advanced fluid-handling solutions for large number of analytical instrumentation and in vitro diagnostic systems. It can be made use in high performance liquid chromatographic (HPLC) systems and is quite accurate and precise in injecting 1 μL-5 mL concentration of samples.
Other Notes
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Note: Use VESPEL seals to pH 10, Tefzel seals to pH 14.
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)