Merck
56728
Stainless Steel 1/16 in. Capillary Tubing
L × O.D. × I.D. 30 cm × 1/16 in. × 0.020 in.
Informacje o tej pozycji
eCl@ss:
32119290
UNSPSC Code:
41115700
NACRES:
NB.35
Pomoc techniczna
Pozwól nam pomóc
L × O.D. × I.D.
30 cm × 1/16 in. × 0.020 in.
color
yellow band
General description
316 stainless steel, 1/16 in. O.D. One piece each. The 5 cm, 10 cm and 20 cm lengths are straight, all other dimensions are coiled
Other Notes
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)