Przejdź do zawartości
55044
RheBuild® Kit
for use with 7000/7010
Informacje o tej pozycji
UNSPSC Code:
41115854
Pomoc techniczna
Pozwól nam pomóc
compatibility
for use with 7000/7010
General description
These kits include all of the tools and parts you need to repair your Rheodyne injector. The parts included are those most likely to be damaged, worn, or lost. Kits for front-loading injectors include: rotor seal, stator face assembly, isolation seal, needle guide, needle port cleaner, 2 hex keys, operating instructions, and a mini-manual. Parts may vary, depending on valve model.
Other Notes
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
RheBuild is a registered trademark of IDEX Health & Science, LLC
Still not finding the right product?
Explore all of our products under RheBuild® Kit
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)