Merck
27401
Screw cap
black polypropylene hole cap,
large opening in top of cap, no septum
, pkg of 100 ea, thread for 8-425
Informacje o tej pozycji
UNSPSC Code:
41122001
Pomoc techniczna
Pozwól nam pomóc
material
black polypropylene hole cap
packaging
pkg of 100 ea
fitting
thread for 8-425
application(s)
general analytical
General description
Screw caps (large, opening in top of cap, no septum, thread 8-425) are available as hole caps for use with sealing septa. Polypropylene caps can withstand a maximum temperature of 135°C.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)