Merck
24846
Battery charger for M-5 / M-30
AC/DC input 220 V AC
Informacje o tej pozycji
UNSPSC Code:
41104008
NACRES:
NB.21
Pomoc techniczna
Pozwól nam pomóc
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
General description
Battery charger for M-5 / M-30 are used as accessories in flow calibration devices for air sampling pumps.
Still not finding the right product?
Explore all of our products under Battery charger for M-5 / M-30
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)