Merck
24844
Battery charger for M-5 / M-30
AC/DC input 110 V AC
Informacje o tej pozycji
UNSPSC Code:
41104008
Pomoc techniczna
Pozwól nam pomóc
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
General description
Battery charger for M-5 / M-30 is used as accessories in flow calibration devices for air sampling pumps employed in air monitoring.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)