Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
eCl@ss:
32049213
UNSPSC Code:
41115700
NACRES:
SB.54
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóctechnique(s)
gas chromatography (GC): suitable using :Suitable
General description
The Merlin Microseal System is a septum-less system that provides very long life. Because there is not a septum to pierce, septum fragments will not be formed and deposited in the inlet liner. The septum contains a series of seals (wiper rib, o-rings, and a duckbill valve) that allow a needle to enter while maintaining a leak-free seal. The septum can be used only with a syringe that has a 23 gauge "blunt tipped" needle, or with an SPME fiber assembly with a 23 gauge needle. The thicker shaft is required to make the necessary contact with the septum seals.
Other Notes
Three septa versions are available:
Note: Do not use with beveled tips.
- Low Pressure for use with 23 gauge syringe needles, and injection port pressures between 1 and 45 psi. Do not use with syringe needles that have beveled tips.
- General Purpose for use with 23 gauge syringe needles, and injection port pressures between 3 and 100 psi. Do not use with syringe needles that have beveled tips.
- SPME for use with 23 gauge SPME fiber assemblies. Do not use with SPME fiber assemblies that have beveled tips.
Note: Do not use with beveled tips.
Legal Information
MicroSeal is a trademark of Merlin Instrument Co.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description 1 Low Pressure (1-45 psi) septum | description Plain, joint: ST/NS 24/40, PTFE stopcock | description capacity 250 mL, PTFE stopcock | description capacity 500 mL, PTFE stopcock |
| technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) - | technique(s) - | technique(s) - |
Still not finding the right product?
Explore all of our products under Merlin Microseal™ System Replacement Septum
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?