Merck
22386
Supeltex M-1 Packed Column Ferrule, 1/16 in. Column O.D.
I.D. 1/16 in., 1/16 in. O.D. Column, pkg of 10 ea
Informacje o tej pozycji
UNSPSC Code:
41121815
NACRES:
SB.54
Pomoc techniczna
Pozwól nam pomóc
packaging
pkg of 10 ea
technique(s)
gas chromatography (GC): suitable using :Suitable
I.D.
1/16 in.
fitting
1/16 in. ferrule for 1/16 in. O.D. packed column
compatibility
1/16 in. O.D. Column
General description
Characteristics: Ideal for connection to mass spectrometers. High reusability. Isothermal use only.
Material Composition: Ceramic-filled PTFE
Maximum Temperature:
250 °C
Legal Information
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)