Merck
21992
Static Dilution Bottle w/ Mininert® Valve
Informacje o tej pozycji
UNSPSC Code:
41104008
NACRES:
NB.21
Pomoc techniczna
Pozwól nam pomóc
packaging
pkg of 1 bottle
Quality Segment
technique(s)
active air sampling: suitable
flask volume
2 L
application(s)
air monitoring
environmental
industrial hygiene
General description
  • Static dilution bottle w/ Mininert® valve features a two liter round bottomed flask with a threaded neck.
  • Use the static dilution bottle to prepare gaseous volatile organic standards, using a technique developed by the US EPA for air analyses.
Features and Benefits
MinInert valve included
Legal Information
Mininert is a registered trademark of Valco Instruments Co. Inc.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)